Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lst-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383241.1 | 3 - 4 | 423 | 71 | ||
NM_001383242.1 | 4 - 5 | 1064 | 71 | |||
NM_182290.5 | NP_872090.1 | 3 - 4 | 427 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |