Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nuo-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071332.6 | NP_503733.1 | 2 - 3 | 851 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |