Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-94 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383341.1 | 5 - 6 | 1094 | 93 | ||
NM_001383342.1 | 4 - 5 | 1061 | 93 | |||
GenBank mRNA | AY204182.1 | - | 5 - 6 | 1050 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |