Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001356689.2 | NP_001343632.1 | 8 - 9 | 952 | 83 | |
NM_001383329.1 | 8 - 9 | 824 | 83 | |||
NM_072382.9 | NP_504783.2 | 8 - 9 | 957 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |