Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: exo-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026413.6 | NP_001021584.1 | 1 - 2 | 274 | 79 | |
NM_001026414.4 | NP_001021585.1 | 1 - 2 | 296 | 79 | ||
GenBank mRNA | AF034258.1 | - | 1 - 2 | 268 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |