Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: smu-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060878.6 | NP_493279.1 | 5 - 6 | 848 | 108 | |
GenBank mRNA | AF330595.1 | - | 5 - 6 | 820 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |