Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265202.3 | NP_001252131.1 | 8 - 9 | 1130 | 123 | |
GenBank mRNA | AY652944.1 | - | 7 - 8 | 998 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |