Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mys-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322689.3 | NP_001309531.1 | 3 - 4 | 549 | 71 | |
NM_001330941.4 | NP_001317768.1 | 3 - 4 | 551 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |