Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ero-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001377680.2 | NP_001364667.1 | 3 - 4 | 734 | 104 | |
NM_001377681.1 | NP_001364626.1 | 4 - 5 | 629 | 104 | ||
NM_001377682.2 | NP_001364632.1 | 2 - 3 | 210 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |