Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egl-26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313583.2 | NP_001300512.1 | 3 - 4 | 732 | 71 | |
NM_061251.5 | NP_493652.1 | 3 - 4 | 735 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |