Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Ce02426650_g1
See other ETR-1 GE Assays ›
Gene Symbol:
etr-1
Gene Name:
protein-coding
Gene Aliases:
CELE_T01D1.2
Chromosome Location:
Chr.II: 154166 - 170630 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID
Ce02426650_g1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: etr-1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001381290.2
NP_001367867.1
9 - 10
1454
68
NM_001381291.1
NP_001368521.1
9 - 10
1448
68
NM_001381292.2
NP_001367871.1
10 - 11
1446
68
NM_001381293.1
NP_001367870.1
10 - 11
1551
68
NM_001381294.1
NP_001367872.1
9 - 10
1429
68
NM_001381296.1
NP_001368522.1
8 - 9
1032
68
NM_001381297.1
NP_001368523.1
8 - 9
1026
68
NM_001381298.1
NP_001368524.1
7 - 8
807
68
NM_001381299.1
NP_001368525.1
6 - 7
906
68
NM_001381300.1
NP_001368526.1
6 - 7
900
68
NM_001381301.1
NP_001368527.1
5 - 6
681
68
NM_001381302.1
NP_001367869.1
5 - 6
696
68
NM_001381303.1
NP_001368528.1
5 - 6
708
68
NM_001381304.1
NP_001368529.1
5 - 6
702
68
NM_001381305.1
NP_001368530.1
4 - 5
483
68
NM_001381306.1
NP_001368531.1
3 - 4
480
68
NM_001381307.1
NP_001368532.1
3 - 4
474
68
NM_001381308.1
NP_001368533.1
2 - 3
255
68
NM_001383771.1
8 - 9
1245
68
NR_132573.1
9 - 10
1392
68
GenBank mRNA
EF523766.1
-
9 - 10
1392
68
EF523770.1
-
8 - 9
1179
68
U53931.1
-
9 - 10
1455
68
Back To Top
More Information
Set Membership:
Amplicon length less than or equal to 70
3' Most
Detect Genomic DNA
Recommended Assay
Caenorhabditis elegans
Probe spans exons
Gene Ontology Categories:
Function(s)
Process(es)
regulation of alternative mRNA splicing, via spliceosome
mRNA splice site selection
determination of adult lifespan
nucleic acid binding
RNA binding
mRNA binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-c4z4p:80/10.239.161.116:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0