Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sgcb-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061309.5 | NP_493710.2 | 3 - 4 | 343 | 157 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |