Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxc-40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318282.3 | NP_001305211.1 | 1 - 2 | 53 | 70 | |
NM_061474.7 | NP_493875.1 | 3 - 4 | 374 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |