Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srh-72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061620.2 | NP_494021.1 | 1 - 2 | 371 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |