Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptc-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129178.3 | NP_001122650.1 | 6 - 7 | 888 | 67 | |
NM_061982.7 | NP_494383.2 | 7 - 8 | 987 | 67 | ||
NM_061983.8 | NP_494384.2 | 7 - 8 | 976 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |