Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sqv-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061993.8 | NP_494394.1 | 5 - 6 | 1200 | 104 | |
GenBank mRNA | AY241927.1 | - | 3 - 4 | 926 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Caenorhabditis elegans Probe spans exons |