Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: apd-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062169.6 | NP_494570.1 | 7 - 8 | 1335 | 82 | |
NM_062170.5 | NP_494571.1 | 3 - 4 | 615 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |