Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bcmo-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062293.6 | NP_494694.2 | 2 - 3 | 488 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |