Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srg-56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182416.1 | NP_872216.1 | 1 - 2 | 205 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |