Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: abcx-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062920.4 | NP_495321.3 | 6 - 7 | 1184 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |