Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: agr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026981.6 | NP_001022152.3 | 23 - 24 | 3672 | 65 | |
GenBank mRNA | AM773423.1 | - | 23 - 24 | 3672 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |