Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sqv-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063035.6 | NP_495436.1 | 1 - 2 | 115 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |