Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alfa-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063202.8 | NP_495603.1 | 2 - 3 | 192 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |