Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alfa-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063202.8 | NP_495603.1 | 6 - 7 | 1463 | 79 | |
NM_063203.6 | NP_495604.1 | 5 - 6 | 1468 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |