Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glr-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267197.2 | NP_001254126.1 | 4 - 5 | 670 | 165 | |
NM_001373747.3 | NP_001359784.1 | 4 - 5 | 673 | 165 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |