Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acr-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063246.5 | NP_495647.1 | 7 - 8 | 919 | 110 | |
GenBank mRNA | AF187012.1 | - | 7 - 8 | 919 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |