Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373748.2 | NP_001360447.1 | 8 - 9 | 4116 | 87 | |
NM_063261.5 | NP_495662.2 | 7 - 8 | 3934 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |