Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ltd-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322746.3 | NP_001309573.1 | 7 - 8 | 1332 | 77 | |
NM_063296.6 | NP_495697.2 | 10 - 11 | 2099 | 77 | ||
GenBank mRNA | AF489839.1 | - | 10 - 11 | 2121 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |