Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-105 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129123.3 | NP_001122595.1 | 6 - 7 | 1474 | 179 | |
NM_001267210.3 | NP_001254139.1 | 7 - 8 | 1435 | 179 | ||
NM_001377828.2 | NP_001364812.1 | 7 - 8 | 1462 | 179 | ||
NM_001381517.1 | NP_001368112.1 | 7 - 8 | 1456 | 179 | ||
NM_001381518.2 | NP_001367274.1 | 7 - 8 | 1456 | 179 | ||
NM_063301.6 | NP_495702.1 | 6 - 7 | 1274 | 179 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |