Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctg-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267227.3 | NP_001254156.1 | 4 - 5 | 861 | 103 | |
NM_001381528.1 | NP_001367210.1 | 4 - 5 | 825 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |