Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mel-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026757.4 | NP_001021928.1 | 6 - 7 | 1573 | 59 | |
NM_001383917.1 | 6 - 7 | 1578 | 59 | |||
NM_063593.6 | NP_495994.1 | 6 - 7 | 1579 | 59 | ||
GenBank mRNA | U86640.1 | - | 6 - 7 | 1570 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |