Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mel-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026757.4 | NP_001021928.1 | 10 - 11 | 2482 | 122 | |
NM_001381562.1 | NP_001367038.1 | 9 - 10 | 2172 | 122 | ||
NM_063593.6 | NP_495994.1 | 9 - 10 | 2164 | 122 | ||
GenBank mRNA | U86640.1 | - | 9 - 10 | 2155 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |