Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lec-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276739.3 | NP_001263668.1 | 9 - 10 | 760 | 73 | |
NM_063758.8 | NP_496159.1 | 9 - 10 | 795 | 73 | ||
GenBank mRNA | AB038504.1 | - | 8 - 9 | 702 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |