Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mpz-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026867.4 | NP_001022038.2 | 24 - 25 | 3052 | 92 | |
NM_001083157.4 | NP_001076626.2 | 18 - 19 | 2445 | 92 | ||
NM_001129127.4 | NP_001122599.1 | 17 - 18 | 2391 | 92 | ||
NM_001129129.4 | NP_001122601.1 | 22 - 23 | 2874 | 92 | ||
NM_001267333.3 | NP_001254262.1 | 1 - 2 | 66 | 92 | ||
NM_001377853.2 | NP_001364655.1 | 25 - 26 | 3105 | 92 | ||
NM_001377854.2 | NP_001364657.1 | 24 - 25 | 3018 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |