Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mpz-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026867.4 | NP_001022038.2 | 28 - 29 | 3566 | 72 | |
NM_001083157.4 | NP_001076626.2 | 22 - 23 | 2959 | 72 | ||
NM_001129127.4 | NP_001122599.1 | 21 - 22 | 2905 | 72 | ||
NM_001129129.4 | NP_001122601.1 | 26 - 27 | 3388 | 72 | ||
NM_001267333.3 | NP_001254262.1 | 5 - 6 | 580 | 72 | ||
NM_001377853.2 | NP_001364655.1 | 29 - 30 | 3619 | 72 | ||
NM_001377854.2 | NP_001364657.1 | 28 - 29 | 3532 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |