Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mpz-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026867.4 | NP_001022038.2 | 40 - 41 | 6987 | 83 | |
NM_001083157.4 | NP_001076626.2 | 34 - 35 | 6380 | 83 | ||
NM_001129127.4 | NP_001122599.1 | 33 - 34 | 6326 | 83 | ||
NM_001129129.4 | NP_001122601.1 | 38 - 39 | 6821 | 83 | ||
NM_001267333.3 | NP_001254262.1 | 17 - 18 | 4001 | 83 | ||
NM_001267334.3 | NP_001254263.1 | 10 - 11 | 2714 | 83 | ||
NM_001267335.3 | NP_001254264.1 | 5 - 6 | 1184 | 83 | ||
NM_001377853.2 | NP_001364655.1 | 41 - 42 | 7040 | 83 | ||
NM_001377854.2 | NP_001364657.1 | 40 - 41 | 6965 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |