Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mix-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063930.8 | NP_496331.1 | 9 - 10 | 2058 | 144 | |
GenBank mRNA | U96387.1 | - | 9 - 10 | 1920 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |