Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mdt-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064063.5 | NP_496464.2 | 3 - 4 | 883 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Caenorhabditis elegans Probe spans exons |