Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001377877.2 | NP_001364647.1 | 12 - 13 | 7084 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |