Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kel-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267381.2 | NP_001254310.1 | 1 - 2 | 179 | 71 | |
GenBank mRNA | AB023424.1 | - | 1 - 2 | 171 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |