Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129177.4 | NP_001122649.1 | 3 - 4 | 524 | 65 | |
NM_064144.6 | NP_496545.1 | 3 - 4 | 539 | 65 | ||
GenBank mRNA | L39212.1 | - | 3 - 4 | 548 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |