Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ash-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001330962.4 | NP_001317781.1 | 3 - 4 | 499 | 72 | |
NM_001373798.2 | NP_001360415.1 | 3 | 504 | 72 | ||
NM_001373799.3 | NP_001359515.1 | 3 - 4 | 543 | 72 | ||
NM_064154.3 | NP_496555.1 | 3 - 4 | 500 | 72 | ||
NR_138517.1 | 3 - 4 | 1131 | 72 | |||
GenBank mRNA | EU178102.1 | - | 3 - 4 | 488 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |