Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glb-29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267402.3 | NP_001254331.1 | 8 - 9 | 900 | 72 | |
NM_001267403.3 | NP_001254332.1 | 8 - 9 | 927 | 72 | ||
NM_001267404.4 | NP_001254333.1 | 6 - 7 | 678 | 72 | ||
NM_001267405.2 | NP_001254334.1 | 4 - 5 | 431 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |