Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cnt-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027242.3 | NP_001022413.1 | 2 - 3 | 668 | 74 | |
NM_001381624.1 | NP_001368630.1 | 2 - 3 | 164 | 74 | ||
NM_001383965.1 | 7 - 8 | 924 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |