Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: set-24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064178.6 | NP_496579.3 | 1 - 2 | 242 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |