Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rgs-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267421.4 | NP_001254350.3 | 5 - 6 | 592 | 70 | |
NM_001373807.2 | NP_001360477.1 | 5 - 6 | 627 | 70 | ||
NM_001377886.2 | NP_001364555.1 | 7 - 8 | 801 | 70 | ||
NM_001381631.1 | NP_001368098.1 | 4 - 5 | 400 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |