Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zyx-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267435.3 | NP_001254364.1 | 3 - 4 | 415 | 132 | |
NM_064375.8 | NP_496776.1 | 4 - 5 | 667 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |