Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gur-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064546.5 | NP_496947.3 | 3 - 4 | 583 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |