Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129192.5 | NP_001122664.2 | 2 - 3 | 357 | 95 | |
NM_064581.4 | NP_496982.2 | 4 - 5 | 681 | 95 |
Gene Symbol: Y54G11A.9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383990.1 | 2 - 3 | 246 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |