Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trip-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064699.5 | NP_497100.1 | 2 - 3 | 658 | 78 | |
GenBank mRNA | AF197575.1 | - | 2 - 3 | 668 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |